Learn More
IBA Lifesciences™ CD3/CD28 Streptamer™ Kit, Mouse
Shop All IBA Lifesciences ProductsDescription
The CD3/CD28 Streptamer™ approach is based on our Strep-tag™ technology and facilitates T cell activation and expansion. Low affinity, Fab fragments fused to a Twin-Strep-tag™ (Fab-Streps) against CD3 and CD28 are multimerized on a Strep-Tactin™ backbone. The resulting CD3/CD28 Streptamer™ complex stimulates the T cell receptor/CD3 complex (TCR/CD3; signal 1) and the co-stimulatory molecule CD28 (signal 2). Providing these two signals leads to efficient T cell activation and proliferation.
For in vitro stimulation and activation, the soluble CD3/CD28 Streptamers are added to T cells. The addition of biotin reverses the multimerization and causes the dissociation of low affinity Fab fragments from the cells. Consequently, the termination of stimulation is possible at any point in time.
- Reversible reagents due to low affinity Fab fragments
- Accurately defined termination of stimulation
Specifications
Specifications
| Cell Type | Primary Murine T Cells |
| Target Species | Murine |
| For Use With (Application) | T Cell Activation and Expansion |
| Sufficient For | 5 x 107 Total Cells |
| Quantity | 1 Kit |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.