missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD28, Rabbit anti-Rat, Clone: JJ316, Novus Biologicals™
Rabbit Monoclonal Antibody
£207.00 - £454.00
Specifications
| Antigen | CD28 |
|---|---|
| Clone | JJ316 |
| Concentration | 1 mg/mL |
| Dilution | Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Functional, Block/Neutralize |
| Applications | Flow Cytometry, Immunohistochemistry, Immunofluorescence, Functional Assay, Blocking Assay |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18287708
|
Novus Biologicals
NBP2-81052-0.025MG |
0.025 mg |
£207.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18253339
|
Novus Biologicals
NBP2-81052-0.2MG |
0.2 mg |
£454.00
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD28 Monoclonal specifically detects CD28 in Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Functional, Block/Neutralize.Specifications
| CD28 | |
| 1 mg/mL | |
| Flow Cytometry, Immunohistochemistry, Immunofluorescence, Functional Assay, Blocking Assay | |
| Unconjugated | |
| Rabbit | |
| Adaptive Immunity, Apoptosis, Immunology | |
| PBS | |
| 940 | |
| Made by immunizing BALB/c mice with rat CD28 cDNA-transfected A20/J cells. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| JJ316 | |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Functional, Block/Neutralize | |
| Monoclonal | |
| Purified | |
| RUO | |
| Rat | |
| CD28 antigen, CD28 antigen (Tp44), CD28 molecule, MGC138290, T-cell-specific surface glycoprotein CD28, Tp44 | |
| CD28 | |
| IgG κ | |
| Protein A purified | |
| Recognises rat CD28 and can trigger T-cell proliferation in vitro and in vivo. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title