missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD28, Rabbit anti-Rat, Clone: JJ316, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-81052-0.025MG
This item is not returnable.
View return policy
Description
CD28 Monoclonal specifically detects CD28 in Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Functional, Block/Neutralize.
Specifications
| CD28 | |
| Monoclonal | |
| 1 mg/mL | |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Functional, Block/Neutralize | |
| CD28 antigen, CD28 antigen (Tp44), CD28 molecule, MGC138290, T-cell-specific surface glycoprotein CD28, Tp44 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Recognises rat CD28 and can trigger T-cell proliferation in vitro and in vivo. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG κ |
| Flow Cytometry, Immunohistochemistry, Immunofluorescence, Functional Assay, Blocking Assay | |
| JJ316 | |
| Unconjugated | |
| PBS | |
| CD28 | |
| Made by immunizing BALB/c mice with rat CD28 cDNA-transfected A20/J cells. | |
| 0.025 mg | |
| Adaptive Immunity, Apoptosis, Immunology | |
| 940 | |
| Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction