missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD28 Golden Hamster, Clone: 37.51, enQuireBio™
Golden Hamster Monoclonal Antibody
Brand: EnquireBio QAB99-500ug
Additional Details : Weight : 0.01000kg
Specifications
CD28 | |
Monoclonal | |
0.5mg/mL | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2 | |
Cd28 | |
Protein A/G Purified | |
RUO | |
12487 | |
IgG |
Flow Cytometry, Immunoprecipitation | |
37.51 | |
Unconjugated | |
NP_031668 | |
Golden Syrian Hamster | |
500 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |