missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD28 Golden Hamster, Clone: 37.51, enQuireBio™
Golden Hamster Monoclonal Antibody
£62.45 - £191.00
Specifications
Antigen | CD28 |
---|---|
Clone | 37.51 |
Concentration | 0.5mg/mL |
Applications | Flow Cytometry, Immunoprecipitation |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15942668
|
EnquireBio
QAB99-25UG |
25 μg |
£62.45
25µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15932668
|
EnquireBio
QAB99-100UG |
100 μg |
£99.15
100µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15952668
|
EnquireBio
QAB99-500UG |
500 μg |
£191.00
500µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
CD28 | |
0.5mg/mL | |
Monoclonal | |
Golden Syrian Hamster | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2 | |
12487 | |
IgG | |
Protein A/G Purified |
37.51 | |
Flow Cytometry, Immunoprecipitation | |
Unconjugated | |
RUO | |
NP_031668 | |
Cd28 | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |