missing translation for 'onlineSavingsMsg'
Learn More

CD28 Golden Hamster, Clone: 37.51, enQuireBio™

Golden Hamster Monoclonal Antibody

Brand:  EnquireBio QAB99-25ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15942668

  • £62.45 / 25µg
Estimated Shipment: 20-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

CD28
Monoclonal
0.5mg/mL
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2
Cd28
Protein A/G Purified
RUO
12487
IgG
Flow Cytometry, Immunoprecipitation
37.51
Unconjugated
NP_031668
Golden Syrian Hamster
25 μg
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers