missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD279 Armenian Hamster anti-Mouse, BB700, Clone: J43, BD Horizon™
Armenian Hamster Monoclonal Antibody
£140.80 - £349.14
Specifications
| Antigen | CD279 |
|---|---|
| Clone | J43 |
| Concentration | 0.2 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15845119
|
BD Biosciences
566515 |
25 μg |
£147.00 £140.80 / Each Save £6.20 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15835119
|
BD Biosciences
566514 |
0.1 mg |
£364.00 £349.14 / Each Save £14.86 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descripción
CD279 Monoclonal antibody specifically detects CD279 in Mouse samples. It is validated for Flow Cytometry.Especificaciones
| CD279 | |
| 0.2 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| IgG2 κ | |
| Refrigerate (2 to 8°C) |
| J43 | |
| Flow Cytometry | |
| Brilliant Blue 700 (BB700) | |
| Armenian Hamster | |
| Mouse | |
| Primary |
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto