missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD279 Armenian Hamster anti-Mouse, BB700, Clone: J43, BD Horizon™
Armenian Hamster Monoclonal Antibody
Brand: BD Biosciences 566514
This item is not returnable.
View return policy
Description
CD279 Monoclonal antibody specifically detects CD279 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| CD279 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Armenian Hamster | |
| RUO | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| J43 | |
| Brilliant Blue 700 (BB700) | |
| 0.1 mg | |
| Primary | |
| Refrigerate (2 to 8°C) | |
| IgG2 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction