Learn More
CD273 (B7-DC) Monoclonal Antibody (TY25), Functional Grade, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Brand: Affymetrix eBioscience 16-5986-025
Description
The TY25 MAb reacts with mouse B7-DC/PD-L2. B7-DC, a member of the B7 family, has a predicted molecular weight of ∽25 kDa and belongs to the Ig superfamily. The mouse B7-DC has a short cytoplasmic tail (4aa). B7-DC is primarily expressed by sub-populations of dendritic cells, monocytes and macrophages. Although B7-DC has structural and sequence similarities to the B7 family, it does not bind CD28/CTLA-4, but binds PD-1. The interactions between PD-1 and B7-DC/PD-L2 have been reported to be involved in costimulation or suppression of T cell proliferation depending on state of cellular activation. TY25 is a useful tool to study the exact function of B7-DC/PD-L2 in APC/T cell interaction and to characterize the expression pattern of this molecule in mouse.
CD273 (B7-DC) Monoclonal antibody specifically detects CD273 (B7-DC) in Mouse samples. It is validated for Flow Cytometry, Functional Assay, Neutralization
Specifications
| CD273 (B7-DC) | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative; pH 7.2 | |
| Q9WUL5 | |
| Rat | |
| 25 mg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a κ |
| Flow Cytometry, Functional Assay, Neutralization | |
| TY25 | |
| Functional Grade | |
| PDCD1LG2 | |
| B7 dendritic cell molecule; B7DC; B7-DC; bA574F11.2; Btdc; Butyrophilin B7-DC; butyrophilin-like protein; CD273; F730015O22Rik; MGC124039; MGC124040; PD1 ligand 2; PD-1 ligand 2; PD-1-ligand 2; PDCD1 ligand 2; PDCD1L2; PDCD1LG2; Pdl2; PD-L2; Programmed cell death 1 ligand 2; programmed death ligand 2 | |
| Affinity chromatography | |
| RUO | |
| 58205 | |
| 4° C | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.