Learn More
Invitrogen™ CD271 (NGF Receptor) Monoclonal Antibody (ME20.4), eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£92.00 - £3115.00
Specifications
| Antigen | CD271 (NGF Receptor) |
|---|---|
| Clone | ME20.4 |
| Concentration | 0.5 mg/mL |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15537596
|
Invitrogen™
14-9400-80 |
25 μg |
£92.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17120742
|
Invitrogen™
14-9400-37 |
2 mg |
£1376.00
2mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17130742
|
Affymetrix eBioscience
14-9400-95 |
10 mg | |||||||
Description
The MAb ME20.4 recognizes CD271, a 75 kDa type I transmembrane protein that belongs to the low-affinity neurotrophin receptor and tumor necrosis factor receptor superfamily. CD271 is expressed by neurons, Schwann cells, melanocytes, mesenchymal stem/stromal cells, neural crest stem cells, astrocytes, oligodendrocytes, and a subset of lymphoid cells including follicular dendritic cells. CD271 binds to the ligands NGF, BDNF, NTF3, and NTF4. CD271 contains a death domain which induces apoptosis and regulates cell growth and proliferation through interactions with TRAF2, TRAF4, TRAF6, PTPN13, and RANBP9. The ME20.4 MAb recognizes human, Non-human Primate, mouse, rat, sheep, rabbit, cat, dog, and pig CD271.
Neurotrophic factors control the survival, differentiation and maintenance of neurons in the peripheral and central nervous systems, and of other neural crest-derived cell types. Developing sympathetic neurons are absolutely dependent upon nerve growth factor (NGF) during the period of target competition in vivo. Antibodies reacting specifically with NGFR p75 are useful tools in the detection and characterization of NGFR p75, to enhance our understanding of a wide range of phenomena in the development, plasticity and repair of the nervous system.Specifications
| CD271 (NGF Receptor) | |
| 0.5 mg/mL | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat, Canine, Rabbit, Monkey, Sheep, Pig | |
| P07174, P08138, Q9Z0W1 | |
| IgG1 κ | |
| Affinity chromatography | |
| Antibody |
| ME20.4 | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| 100349095, 100525607, 101119064, 18053, 24596, 4804, 491071 | |
| Primary | |
| 4°C | |
| Ngfr |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.