missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD267 (TACI) Rat, anti-Mouse, APC, Clone: 8F10, STEMCELL Technologies
Rat Monoclonal Antibody
£176.85 - £322.65
Specifications
| Antigen | CD267 (TACI) |
|---|---|
| Clone | 8F10 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | APC |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17109141
|
Stemcell Technologies
60116AZ.1 |
50 μg |
£176.85
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17199131
|
Stemcell Technologies
60116AZ |
100 μg |
£322.65
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- The 8F10 antibody reacts with mouse CD267 (transmembrane activator and CAML interactor, or TACI), an ∼30 kDa type III transmembrane protein of the TNF receptor superfamily that is expressed primarily on the surface of maturing subsets of splenic B cells, including transition type 2 (T2) and marginal zone (MZ) B cells, as well as on plasma cells and plasmablasts
- Expression of CD267 is upregulated on B cells exposed to LPS both in vitro and in vivo
- CD267 plays important roles in humoral immunity by regulating B cell survival, proliferation, and maturation through its interaction with the ligands APRIL and BAFF
- Ligand binding stimulates activation of several transcription factors, including NF-kappa-B, and has been shown to induce either proliferation or apoptosis, suggesting a dual role in the regulation of B cells
- CD267 also appears to have an essential function in immunoglobulin isotype switching and B cell differentiation in response to thymus-independent antigens
Specifications
| CD267 (TACI) | |
| Flow Cytometry | |
| APC | |
| Mouse | |
| Primary |
| 8F10 | |
| Monoclonal | |
| Rat | |
| IgG2a κ |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title