missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD25 Mouse anti-Human, Brilliant Ultraviolet 737, Clone: 2A3, BD Horizon™
Mouse Monoclonal Antibody
£177.56 - £402.06
Specifications
| Antigen | CD25 |
|---|---|
| Clone | 2A3 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | Brilliant Ultraviolet 737 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17207904
|
BD Biosciences
612807 |
25 Tests |
£180.00 £177.56 / 25 tests Save £2.44 1% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16393528
|
BD Biosciences
612806 |
100 Tests |
£410.00 £402.06 / 100 tests Save £7.94 1% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD25 Monoclonal antibody specifically detects CD25 in Human samples. It is validated for Flow Cytometry.Specifications
| CD25 | |
| Flow Cytometry | |
| Brilliant Ultraviolet 737 | |
| RUO | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| IgG1 κ | |
| Affinity Purified |
| 2A3 | |
| Monoclonal | |
| Mouse | |
| Human | |
| Human Phytohemagglutinin-activated T Cells | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title