missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD25 Mouse anti-Human, Brilliant Ultraviolet 737, Clone: 2A3, BD Horizon™
Mouse Monoclonal Antibody
Brand: BD Biosciences 612806
This item is not returnable.
View return policy
Description
CD25 Monoclonal antibody specifically detects CD25 in Human samples. It is validated for Flow Cytometry.
Specifications
| CD25 | |
| Monoclonal | |
| Brilliant Ultraviolet 737 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG1 κ |
| Flow Cytometry | |
| 2A3 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Human Phytohemagglutinin-activated T Cells | |
| 100 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction