missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD25/ IL2RA (Activated Lymphocyte Marker) Mouse, Clone: IL2RA/2395, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 3559-MSM5-P0
Additional Details : Weight : 0.01000kg
Specifications
CD25/ IL2RA (Activated Lymphocyte Marker) | |
Cell Surface and Cytoplasmic | |
IL2RA/2395 | |
Unconjugated | |
Mouse | |
55kDa | |
Protein A/G purified from Bioreactor concentrate. | |
RUO | |
3559 | |
IgG2b |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
Recombinantfragment of human IL2RA protein (apprx. aa 42-183) | |
PHA-stimulated human lymphocytes. Human lymph nodeor tonsil | |
20 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |