missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD25/ IL2RA (Activated Lymphocyte Marker) Mouse, Clone: IL2RA/2395, enQuireBio™
Mouse Monoclonal Antibody
£181.83 - £395.00
Specifications
Antigen | CD25/ IL2RA (Activated Lymphocyte Marker) |
---|---|
Clone | IL2RA/2395 |
Concentration | 200μg/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15960268
|
EnquireBio
3559-MSM5-P1 |
100 μg |
£395.00
100µg |
Estimated Shipment: 12-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15950268
|
EnquireBio
3559-MSM5-P0 |
20 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
CD25/ IL2RA (Activated Lymphocyte Marker) | |
200μg/mL | |
Monoclonal | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
Recombinantfragment of human IL2RA protein (apprx. aa 42-183) | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
55kDa |
IL2RA/2395 | |
Immunohistochemistry | |
Unconjugated | |
RUO | |
3559 | |
IgG2b | |
Protein A/G purified from Bioreactor concentrate. | |
Cell Surface and Cytoplasmic | |
PHA-stimulated human lymphocytes. Human lymph nodeor tonsil |