missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD249 Recombinant Rabbit Monoclonal Antibody (23GB5325), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | CD249 |
|---|---|
| Clone | 23GB5325 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089550
![]() |
Invitrogen™
MA553886 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088588
|
Invitrogen™
MA553887 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD249 Recombinant Monoclonal antibody specifically detects CD249 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| CD249 | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P16406, P50123, Q07075 | |
| 13809, 2028, 64017 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB5325 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Aminopeptidase A; APA; AP-A; CD249 Antigen; Differentiation Antigen Gp160; EAP; EC 3.4.11; EC 3.4.11.7; ENPEP; Glutamyl Aminopeptidase; Glutamyl Aminopeptidase (Aminopeptidase A); gp160 | |
| A synthesized peptide derived from human Aminopeptidase A/CD249 (900-957AA). | |
| Primary | |
| -20°C | |
| ENPEP |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title