missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD249 Recombinant Rabbit Monoclonal Antibody (23GB5325), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553887
This item is not returnable.
View return policy
Description
CD249 appears to have a role in the catabolic pathway of the renin-angiotensin system. Probably plays a role in regulating growth and differentiation of early B-lineage cells.
Specifications
| CD249 | |
| Recombinant Monoclonal | |
| 1.5 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P16406, P50123, Q07075 | |
| ENPEP | |
| A synthesized peptide derived from human Aminopeptidase A/CD249 (900-957AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry | |
| 23GB5325 | |
| Unconjugated | |
| ENPEP | |
| Aminopeptidase A; APA; AP-A; CD249 Antigen; Differentiation Antigen Gp160; EAP; EC 3.4.11; EC 3.4.11.7; ENPEP; Glutamyl Aminopeptidase; Glutamyl Aminopeptidase (Aminopeptidase A); gp160 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 13809, 2028, 64017 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction