missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD24 Antibody (SR1522), Novus Biologicals™
Rabbit Monoclonal Antibody
£282.00 - £367.00
Specifications
| Antigen | CD24 |
|---|---|
| Clone | SR1522 |
| Dilution | Western Blot 1:500-1:2000, Immunoprecipitation 1:50 |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18680834
|
Novus Biologicals
NBP3-22448-50ul |
50 μg |
£282.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18622575
|
Novus Biologicals
NBP3-22448-100ul |
100 μL |
£367.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD24 Monoclonal antibody specifically detects CD24 in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
| CD24 | |
| Western Blot 1:500-1:2000, Immunoprecipitation 1:50 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 100133941 | |
| Affinity purified |
| SR1522 | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| B Cell Development and Differentiation Markers, Cancer, Neuronal Cell Markers, Stem Cell Markers | |
| CD 24, CD24 antigen, CD24 antigen (small cell lung carcinoma cluster 4 antigen), CD24 molecule, CD24Asignal transducer CD24, FLJ22950, FLJ43543, MGC75043, Small cell lung carcinoma cluster 4 antigen | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title