missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD24 Antibody (SR1522), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22448-100ul
This item is not returnable.
View return policy
Description
CD24 Monoclonal antibody specifically detects CD24 in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| CD24 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| Western Blot, Immunoprecipitation | |
| SR1522 | |
| Western Blot 1:500-1:2000, Immunoprecipitation 1:50 | |
| CD 24, CD24 antigen, CD24 antigen (small cell lung carcinoma cluster 4 antigen), CD24 molecule, CD24Asignal transducer CD24, FLJ22950, FLJ43543, MGC75043, Small cell lung carcinoma cluster 4 antigen | |
| A synthesized peptide derived from human CD24 (Uniprot number: P25063) | |
| 100 μL | |
| B Cell Development and Differentiation Markers, Cancer, Neuronal Cell Markers, Stem Cell Markers | |
| 100133941 | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction