Learn More
Epredia™ Lab Vision™ CD23, Rabbit Monoclonal Antibody
Obtain accurate and reproducible results in immunohistochemistry procedures with Epredia™ CD23, Rabbit Monoclonal Antibody.
Brand: Epredia™ RM-9123-RQ
Code : LD
Additional Details : Weight : 0.02000kg
Description
CD23 is a 45 kDa glycoprotein, which is present on a subpopulation of freshly isolated peripheral blood and tonsil B cells and strongly expressed on EBV-transformed B lymphoblasts. The CD23 molecule is identical to the low affinity IgE receptor found on B cells. Expression of CD23 has been detected in neoplastic cells from cases of B cell chronic lymphocyctic leukaemia and some cases of centroblastic/centrocytic lymphoma.Host Species: Rabbit
Clone: SP23
Isotype: IgG
Species Reactivity: Human. Others not tested.
Epitope: Not known
Immunogen: Recombinant protein encoding human CD23 48-248aa
Molecular Weight: 45 kDa
Positive Control: Tonsil
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
CD23 | |
Monoclonal | |
Unconjugated | |
Recombinant protein encoding human CD23 48-248aa | |
IVD | |
Primary |
Immunohistochemistry (Paraffin) | |
SP23 | |
Rabbit | |
12 mL | |
Oncogenesis | |
Ready-to-Use |