missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Lab Vision™ CD23, Rabbit Monoclonal Antibody
Obtain accurate and reproducible results in immunohistochemistry procedures with Epredia™ CD23, Rabbit Monoclonal Antibody.
£154.02 - £875.00
Specifications
Antigen | CD23 |
---|---|
Clone | SP23 |
Applications | Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
12693777
|
Epredia™
RM-9123-S1 |
Concentrated | 500 μL |
£550.00
500µL |
Estimated Shipment: 03-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12653777
|
Epredia™
RM-9123-S |
Concentrated | 1 mL |
£875.00
1mL |
Estimated Shipment: 03-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12623777
|
Epredia™
RM-9123-R7 |
Prediluted | 7 mL |
£433.00
7mL |
Estimated Shipment: 03-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12673777
|
Epredia™
RM-9123-S0 |
Concentrated | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12643777
|
Epredia™
RM-9123-RQ |
Ready-to-Use | 12 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
CD23 is a 45 kDa glycoprotein, which is present on a subpopulation of freshly isolated peripheral blood and tonsil B cells and strongly expressed on EBV-transformed B lymphoblasts. The CD23 molecule is identical to the low affinity IgE receptor found on B cells. Expression of CD23 has been detected in neoplastic cells from cases of B cell chronic lymphocyctic leukaemia and some cases of centroblastic/centrocytic lymphoma.Host Species: Rabbit
Clone: SP23
Isotype: IgG
Species Reactivity: Human. Others not tested.
Epitope: Not known
Immunogen: Recombinant protein encoding human CD23 48-248aa
Molecular Weight: 45 kDa
Positive Control: Tonsil
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
CD23 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
IVD | |
Recombinant protein encoding human CD23 48-248aa |
SP23 | |
Monoclonal | |
Rabbit | |
Oncogenesis | |
Primary |