Learn More
CD202b (TIE2) Monoclonal Antibody (TEK4), eBioscience™, Invitrogen™

Rat Monoclonal Antibody
£62.50 - £341.00
Specifications
| Antigen | CD202b (TIE2) |
|---|---|
| Clone | TEK4 |
| Concentration | 0.5 mg/mL |
| Applications | Flow Cytometry, Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15277667
|
Invitrogen
14-5987-81 |
50 μg | |||||||
15287667
![]() |
Invitrogen™
14-5987-82 |
100 μg |
£96.75
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15297667
|
Invitrogen™
14-5987-85 |
500 μg |
£341.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Description: The TEK4 monoclonal antibody reacts with mouse Tie-2, also known as CD202. A member of the tyrosine kinase receptor family, Tie-2 is expressed on endothelial and a subset of hematopoietic cells and is believed to play a role in both angiogenesis and hematopoiesis during development of the mouse embryo. In fetal liver and adult bone marrow, Tie-2 is expressed by a subpopulation of hematopoietic progenitor cells characterized as Lineage markers^-, c-Kit^+, Sca1^+ cells. Long-term multilineage repopulating cells were detected in Tie-2^+, Lineage^-, c-Kit^+, Sca1^+ cells but not in Tie-2^-, Lineage^-, c-Kit^+, Sca1^+ cells. Applications Reported: This TEK4 antibody has been reported for use in flow cytometric analysis, and immunohistology staining of frozen tissue sections. Applications Tested: The TEK4 antibody has been tested by blocking of staining with fluorochrome conjugated TEK4 on mouse bone marrow cells. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.
The TEK receptor tyrosine kinase is expressed almost exclusively in endothelial cells. TEK signaling appears to be critical for endothelial cell-smooth muscle cell communication in venous morphogenesis, and defects in TEK are associated with inherited venous malformations. The ligand angiopoietin-1 binds to this receptor and mediates a signaling pathway that functions in embryonic vascular development. Immunoblotting showed that TIE2 expression was increased by thyroid-stimulating hormone and agents that increased intracellular cAMP. HSCs expressing the receptor tyrosine kinase TIE2 are quiescent and antiapoptotic and comprise a side population of HSCs that adhere to osteoblasts in the bone marrow niche.Specifications
| CD202b (TIE2) | |
| 0.5 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| Q02858 | |
| IgG1 κ | |
| Affinity chromatography | |
| Antibody |
| TEK4 | |
| Flow Cytometry, Immunohistochemistry (Frozen) | |
| Unconjugated | |
| Rat | |
| Mouse | |
| 21687 | |
| Primary | |
| 4° C |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.