missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD20 Mouse anti-Human, Clone: 4A7G3, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | CD20 |
|---|---|
| Clone | 4A7G3 |
| Concentration | 1 mg/mL |
| Applications | Western Blot, Flow Cytometry, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16807335
|
Proteintech
60271-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16897325
|
Proteintech
60271-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
mRNA-binding protein involved in proper cytoplasmic distribution of mitochondria. Specifically binds mRNAs of nuclear-encoded mitochondrial proteins in the cytoplasm and regulates transport or translation of these transcripts close to mitochondria, playing a role in mitochondrial biogenesis.Specifications
| CD20 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| B lymphocyte antigen CD20, B1, Bp35, CD20, LEU 16, MS4A1, MS4A1,CD20, MS4A2, S7 | |
| MS4A1 | |
| IgG2b | |
| Protein G | |
| Antibody |
| 4A7G3 | |
| Western Blot, Flow Cytometry, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human | |
| P11836 | |
| 931 | |
| CD20 Fusion Protein Ag17309 | |
| Primary | |
| -20°C | |
| MS4A1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title