missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD20 Mouse anti-Human, Clone: 4A7G3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 60271-1-IG-20UL
This item is not returnable.
View return policy
Description
mRNA-binding protein involved in proper cytoplasmic distribution of mitochondria. Specifically binds mRNAs of nuclear-encoded mitochondrial proteins in the cytoplasm and regulates transport or translation of these transcripts close to mitochondria, playing a role in mitochondrial biogenesis.Specifications
| CD20 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P11836 | |
| MS4A1 | |
| CD20 Fusion Protein Ag17309 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Western Blot, Flow Cytometry, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| 4A7G3 | |
| Unconjugated | |
| MS4A1 | |
| B lymphocyte antigen CD20, B1, Bp35, CD20, LEU 16, MS4A1, MS4A1,CD20, MS4A2, S7 | |
| Mouse | |
| Protein G | |
| RUO | |
| 931 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction