missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD20 Mouse, anti-Human, APC, Clone: 2H7, STEMCELL Technologies
Mouse Monoclonal Antibody
£240.30 - £484.65
Specifications
| Antigen | CD20 |
|---|---|
| Clone | 2H7 |
| Applications | CyTOF, ELISA, Flow Cytometry, Immunohistochemistry, Immunoprecipitation |
| Classification | Monoclonal |
| Conjugate | APC |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17128391
|
Stemcell Technologies
60008AZ.1 |
25 Tests |
£240.30
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17118391
|
Stemcell Technologies
60008AZ |
100 Tests |
£484.65
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Antibody 2H7 reacts with CD20, an ∼35 kDa non-glycosylated type 1 transmembrane protein in the MS4A protein family
- The CD20 polypeptide transverses the cell membrane four times, with only a minor portion of the protein displayed on the cell surface
- The epitope recognized by antibody 2H7 has been mapped to the amino acid sequence YNCEPANPSEKNSPST, located in the large extracellular loop of CD20
- CD20 is expressed on pre-B cells, resting and activated B cells, some follicular dendritic cells, and a subset of T cells
- Expression by B cells is lost upon their differentiation into plasma cells
- By associating with several proteins, including the B cell receptor (CD79), MHC class I and II, CD53, CD81, and CD82, CD20 is involved in initiating intracellular signaling pathways that modulate the activation, proliferation, and differentiation of B cells
- It is thought that CD20 forms multi-subunit ion channels that regulate calcium ion flux across the plasma membrane
Specifications
| CD20 | |
| CyTOF, ELISA, Flow Cytometry, Immunohistochemistry, Immunoprecipitation | |
| APC | |
| Baboon, Cynomolgus, Chimpanzee, Human | |
| Primary |
| 2H7 | |
| Monoclonal | |
| Mouse | |
| IgG2b κ |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title