Learn More
CD20 Mouse, anti-Human, APC, Clone: 2H7, STEMCELL Technologies
Mouse Monoclonal Antibody
Brand: Stemcell Technologies 60008AZ.1
This item is not returnable.
View return policy
Description
- Antibody 2H7 reacts with CD20, an ∼35 kDa non-glycosylated type 1 transmembrane protein in the MS4A protein family
- The CD20 polypeptide transverses the cell membrane four times, with only a minor portion of the protein displayed on the cell surface
- The epitope recognized by antibody 2H7 has been mapped to the amino acid sequence YNCEPANPSEKNSPST, located in the large extracellular loop of CD20
- CD20 is expressed on pre-B cells, resting and activated B cells, some follicular dendritic cells, and a subset of T cells
- Expression by B cells is lost upon their differentiation into plasma cells
- By associating with several proteins, including the B cell receptor (CD79), MHC class I and II, CD53, CD81, and CD82, CD20 is involved in initiating intracellular signaling pathways that modulate the activation, proliferation, and differentiation of B cells
- It is thought that CD20 forms multi-subunit ion channels that regulate calcium ion flux across the plasma membrane
Specifications
| CD20 | |
| Monoclonal | |
| APC | |
| 25 Tests | |
| Baboon, Cynomolgus, Chimpanzee, Human | |
| B Cells |
| CyTOF, ELISA, Flow Cytometry, Immunohistochemistry, Immunoprecipitation | |
| 2H7 | |
| Mouse | |
| Primary | |
| IgG2b κ |