missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD20 Antibody (MS4A1/3410), Novus Biologicals™
Mouse Monoclonal Antibody
£216.00 - £368.00
Specifications
| Antigen | CD20 |
|---|---|
| Clone | MS4A1/3410 |
| Concentration | 0.2 mg/mL |
| Dilution | ELISA, Immunohistochemistry 0.5 - 1 μg/ml, Immunohistochemistry-Paraffin 1-2 μg/ml, Protein Array |
| Applications | ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18663661
|
Novus Biologicals
NBP2-78051-100ug |
100 μg |
£368.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18651991
|
Novus Biologicals
NBP2-78051-20ug |
20 μg |
£216.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD20 Monoclonal antibody specifically detects CD20 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein ArraySpecifications
| CD20 | |
| 0.2 mg/mL | |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array | |
| Unconjugated | |
| Mouse | |
| Adaptive Immunity, B Cell Development and Differentiation Markers, Cancer, Cancer Stem Cells, Cell Biology, Cytokine Research, Immunology, Signal Transduction, Stem Cell Markers, Tumor Biomarkers | |
| 10 mM PBS with 0.05% BSA | |
| B1, B-lymphocyte antigen CD20, B-lymphocyte cell-surface antigen B1, B-lymphocyte surface antigen B1, Bp35MGC3969, CD20 antigen, CD20 receptor, CD20S7, CVID5, LEU-16, Leukocyte surface antigen Leu-16, Membrane-spanning 4-domains subfamily A member 1, membrane-spanning 4-domains, subfamily A, member 1, MS4A2 | |
| A recombinant fragment (around aa 213-297) of human MS4A1 protein (exact sequence is proprietary) (Uniprot: P11836) | |
| Primary | |
| Store at 4°C. |
| MS4A1/3410 | |
| ELISA, Immunohistochemistry 0.5 - 1 μg/ml, Immunohistochemistry-Paraffin 1-2 μg/ml, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P11836, P11836 | |
| 931 | |
| IgG1 κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title