missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD20 Antibody (MS4A1/3410), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-78051-20ug
This item is not returnable.
View return policy
Description
CD20 Monoclonal antibody specifically detects CD20 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array
Specifications
| CD20 | |
| Monoclonal | |
| 0.2 mg/mL | |
| ELISA, Immunohistochemistry 0.5 - 1 μg/ml, Immunohistochemistry-Paraffin 1-2 μg/ml, Protein Array | |
| P11836, P11836 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array | |
| MS4A1/3410 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| B1, B-lymphocyte antigen CD20, B-lymphocyte cell-surface antigen B1, B-lymphocyte surface antigen B1, Bp35MGC3969, CD20 antigen, CD20 receptor, CD20S7, CVID5, LEU-16, Leukocyte surface antigen Leu-16, Membrane-spanning 4-domains subfamily A member 1, membrane-spanning 4-domains, subfamily A, member 1, MS4A2 | |
| A recombinant fragment (around aa 213-297) of human MS4A1 protein (exact sequence is proprietary) (Uniprot: P11836) | |
| 20 μg | |
| Adaptive Immunity, B Cell Development and Differentiation Markers, Cancer, Cancer Stem Cells, Cell Biology, Cytokine Research, Immunology, Signal Transduction, Stem Cell Markers, Tumor Biomarkers | |
| 931 | |
| Store at 4°C. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction