missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD2 Mouse, anti-Human, APC, Clone: RPA-2.10, STEMCELL Technologies
Mouse Monoclonal Antibody
POA - £480.60
Specifications
| Antigen | CD2 |
|---|---|
| Clone | RPA-2.10 |
| Applications | Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | APC |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17138381
|
Stemcell Technologies
60007AZ.1 |
25 Tests |
£207.90
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17128381
|
Stemcell Technologies
60007AZ |
100 Tests |
£480.60
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17107601
|
Stemcell Technologies
100-0432 |
500 Tests | |||||||
Description
- The RPA-2.10 antibody reacts with CD2, an ∼50 kDa type I transmembrane glycoprotein and a member of the immunoglobulin (Ig) superfamily; two Ig-like domains are located in its extracellular portion
- CD2 is broadly expressed on peripheral T and NK cells, dendritic cells, erythrocytes, most thymic T cells, subsets of thymic B cells, and on the endothelium
- CD2 expression appears early during T cell differentiation
- Aberrant expression has been observed in some lymphomas and myeloid leukemias
- CD2 is critically important for T cell activation and signaling, as well as lymphocyte adhesion
- The primary ligand for CD2 is CD58 (LFA-3) located on antigen-presenting cells, with additional ligands comprising CD15 (SSEA-1), CD48 and CD59
- Notably, the RPA-2.10 antibody blocks the mixed lymphocyte reaction
Specifications
| CD2 | |
| Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| APC | |
| Primary |
| RPA-2.10 | |
| Monoclonal | |
| Mouse |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title