Learn More
CD1a Mouse anti-Human, APC, Clone: HI149, BD
Mouse Monoclonal Antibody
£62.31 - £215.63
Specifications
| Antigen | CD1a |
|---|---|
| Clone | HI149 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | APC |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15801309
|
BD Biosciences
561755 |
25 Tests |
£64.20 £62.31 / Each Save £1.89 2% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15899768
|
BD Biosciences
559775 |
100 Tests |
£225.00 £215.63 / Each Save £9.37 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The HI149 monoclonal antibody specifically binds to CD1a. CD1a is a type I transmembrane glycoprotein. The 49 kDa CD1a polypeptide is associated with β2-microglobulin. CD1a is expressed on cortical thymocytes, dendritic cells and Langerhans cells. CD1a has structural similarities to the MHC class I antigen, and plays a role in antigen presentation.
Specifications
| CD1a | |
| Flow Cytometry | |
| APC | |
| RUO | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| HI149 | |
| Monoclonal | |
| Mouse | |
| Human | |
| IgG1 κ | |
| Affinity Purified |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.