Learn More
CD1a Mouse anti-Human, APC, Clone: HI149, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 559775
Description
The HI149 monoclonal antibody specifically binds to CD1a. CD1a is a type I transmembrane glycoprotein. The 49 kDa CD1a polypeptide is associated with β2-microglobulin. CD1a is expressed on cortical thymocytes, dendritic cells and Langerhans cells. CD1a has structural similarities to the MHC class I antigen, and plays a role in antigen presentation.
Specifications
| CD1a | |
| Monoclonal | |
| APC | |
| Mouse | |
| 100 Tests | |
| Immunology | |
| Human | |
| IgG1 κ |
| Flow Cytometry | |
| HI149 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.