missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD196 (CCR6) Monoclonal Antibody (R6H1), PerCP-eFluor™ 710, eBioscience™
GREENER_CHOICE

Mouse Monoclonal Antibody

£316.00

Specifications

Antigen CD196 (CCR6)
Clone R6H1
Concentration 5 μL/Test
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Applications Flow Cytometry
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15539216
GREENER_CHOICE
View Documents
Invitrogen™
46-1969-42
100 Tests
£316.00
100 tests
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Description: This R6H1 monoclonal antibody reacts with CD196 (also known as CCR6), a seven transmembrane G protein-coupled receptor expressed on T, B, dendritic, natural killer, and Langerhans cells. This CC chemokine receptor uniquely binds MIP-3a/CCL20, a chemoattractant for dendritic cells, effector/memory T cells, and B cells. CD196 is also involved in host defense and inflammation at epithelial sites. Furthermore, this receptor has been implicated in Th17 differentiation and CD4+FoxP3+ regulatory T cell development. Applications Reported: This R6H1 antibody has been reported for use in flow cytometric analysis. Applications Tested: This R6H1 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. PerCP-eFluor™ 710 can be used in place of PE-Cy5, PE-Cy5.5 or PerCP-Cy5.5. PerCP-eFluor™ 710 emits at 710 nm and is excited with the blue laser (488 nm). Please make sure that your instrument is capable of detecting this fluorochrome. For a filter configuration, we recommend using the 685 LP dichroic mirror and 710/40 band pass filter, however the 695/40 band pass filter is an acceptable alternative. Our testing indicates that PerCP-eFluor...

CD196, also known as CCR6, is a member of the beta chemokine receptor family and is characterized by a seven transmembrane structure similar to G protein-coupled receptors. This receptor uniquely binds to macrophage inflammatory protein 3 alpha (MIP-3alpha/CCL20), which acts as a chemoattractant for dendritic cells, effector/memory T cells, and B cells. CCR6 is preferentially expressed by immature dendritic cells and memory T cells, playing a crucial role in host defense and inflammation at epithelial sites. It is important for B-lineage maturation and antigen-driven B-cell differentiation, and it regulates the migration and recruitment of dendritic and T cells during inflammatory and immunological responses. Additionally, CCR6 has been implicated in the differentiation of Th17 cells and the development of CD4+FoxP3+ regulatory T cells, highlighting its role in immune regulation. Alternatively spliced transcript variants encoding the same protein have been described for this gene, further emphasizing its functional diversity in immune processes.
Specifications

Specifications

CD196 (CCR6)
5 μL/Test
Flow Cytometry
PerCP-eFluor 710
Mouse
RUO
PBS with BSA and 0.09% sodium azide; pH 7.2
P51684
1235
Primary
Affinity chromatography
R6H1
4°C, store in dark, DO NOT FREEZE!
Monoclonal
Liquid
IgG1 κ
Human
CCR6
BN-1; CC chemokine receptor 6; C-C chemokine receptor type 6; C-C CKR-6; C-C motif chemokine receptor 6; CC-CKR-6; CCR6; CCR-6; CD196; chemokine (C-C motif) receptor 6; chemokine (C-C) receptor 6; chemokine receptor-like 3; CKR6; CKRL3; CKR-L3; Cmkbr6; DCR2; DRY6; DRY-6; G protein-coupled receptor 29; GPR29; GPRCY4; GPR-CY4; G-protein coupled receptor 29; KY411; LARC receptor; seven-transmembrane receptor, lymphocyte, 22; STRL22
CCR6
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.