missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD19 Rat, PE, Clone: 1D3, enQuireBio™
Rat Monoclonal Antibody
£95.92 - £147.00
Specifications
| Antigen | CD19 |
|---|---|
| Clone | 1D3 |
| Concentration | 0.5mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15941768
|
EnquireBio
QAB28-PE-25ug |
25 μg |
£95.92
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15931768
|
EnquireBio
QAB28-PE-100ug |
100 μg |
£147.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
| CD19 | |
| 0.5mg/mL | |
| Monoclonal | |
| Rat | |
| 10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2 | |
| 12478 | |
| IgG2a κ | |
| Protein A/G Purified |
| 1D3 | |
| Flow Cytometry | |
| PE | |
| RUO | |
| NP_033974 | |
| Cd19 | |
| Primary | |
| Store at 2-4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title