Learn More
CD19 Rat, PE, Clone: 1D3, enQuireBio™
Rat Monoclonal Antibody
Brand:  EnquireBio QAB28-PE-100ug
 View more versions of this product
Product Code. 15931768
  • £147.00 / 100µg
Specifications
CD19
Monoclonal
0.5mg/mL
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2
Cd19
Protein A/G Purified
RUO
12478
IgG2a κ
Flow Cytometry
1D3
PE
NP_033974
Rat
100 μg
Primary
Store at 2-4°C.