missing translation for 'onlineSavingsMsg'
Learn More

IBA Lifesciences CD19 Nano-Strep
GREENER_CHOICE

Product Code. 17237610 Shop All IBA Lifesciences Products
Click to view available options
:
Human
This item is not returnable. View return policy

Product Code. 17237610

Brand: IBA Lifesciences 68507150

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Low affinity Nanobody fused to a Twin-Strep-tag™ and specific for murine CD19

  • CD19 Nano-Strep is a recombinantly expressed low affinity nanobody fused to a Twin-Strep-tag™
  • Due to its low affinity for the target, an increase in avidity is required to ensure sufficient binding to the cell surface
  • Combining Nano-Streps with a Strep-Tactin™ backbone leads to the formation of complexes that stably bind to cells.
  • Nano-Streps were developed for Traceless Affinity Cell Selection (Nano-TACS™), which is based on our Strep-tag™ technology

Specifications

Form Lyophilized
For Use With (Application) Fluorescent Cell Staining & Sorting, Magnetic or Affinity Chromatographic Cell Isolation
Species Mouse

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.