missing translation for 'onlineSavingsMsg'
Learn More

CD184 (CXCR4) Mouse, anti-Human, PE, Clone: 12G5, STEMCELL Technologies

Mouse Monoclonal Antibody

£226.80 - £502.20

Specifications

Antigen CD184 (CXCR4)
Clone 12G5
Applications CyTOF, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Functional Assay
Classification Monoclonal
Conjugate PE
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17178961
Encompass_Preferred
View Documents
Stemcell Technologies
60089PE.1
25 Tests
£226.80
25 tests
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17168961
Encompass_Preferred
View Documents
Stemcell Technologies
60089PE
100 Tests
£502.20
100 tests
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • The 12G5 antibody reacts with CD184 (CXCR4 or fusin), an ∼45 kDa seven transmembrane G-protein-coupled receptor expressed on the surface of a broad range of cells, including T and B cells, dendritic cells, monocytes, macrophages, granulocytes, platelets, endothelial and epithelial cells, astrocytes and neurons
  • At least two isoforms of CD184 are generated by alternate mRNA splicing
  • Ligands for CD184 include the CXC chemokine SDF-1 and ubiquitin
  • CD184 has functional roles in mediating chemotaxis, B cell lymphopoiesis and myelopoiesis, cardiogenesis, vascularization and neuronal guidance
  • CD184 acts as a co-receptor for infection by some T cell-tropic isolates of HIV-1 and as an alternative receptor for CD4-independent infection by HIV-2; functions that are blocked by binding of the 12G5 antibody
  • The antibody also partially inhibits SDF-1-induced chemotaxis and calcium influx
  • CD184 is used along with other markers such as CD117 to monitor the emergence of definitive endoderm
Specifications

Specifications

CD184 (CXCR4)
CyTOF, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Functional Assay
PE
African Green Monkey, Baboon, Cynomolgus, Rhesus Monkey, Human
Primary
12G5
Monoclonal
Mouse
IgG2a κ
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.