missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD173 Antibody (BRIC231), Novus Biologicals™
Mouse Monoclonal Antibody
£160.00 - £381.00
Specifications
| Antigen | CD173 |
|---|---|
| Clone | BRIC231 |
| Concentration | 1 mg/mL |
| Dilution | Flow Cytometry 1:10-1:1000, Haemagglutination |
| Applications | Hemagglutination Assay, Flow Cytometry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18088913
|
Novus Biologicals
NB100-62120-0.025MG |
0.025 mg |
£160.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18021064
|
Novus Biologicals
NB100-62120 |
0.1 mg |
£381.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD173 Monoclonal specifically detects CD173 in Human samples. It is validated for Flow Cytometry, Haemagglutination, Immunofluorescence.Specifications
| CD173 | |
| 1 mg/mL | |
| Hemagglutination Assay, Flow Cytometry | |
| Unconjugated | |
| RUO | |
| HEL cell line | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| BRIC231 | |
| Flow Cytometry 1:10-1:1000, Haemagglutination | |
| Monoclonal | |
| Mouse | |
| Human | |
| IgG1 | |
| Affinity Purified | |
| NB100-62120 recognises the type 2 H blood group antigen, recently designated CD173. Active H substances in man, are expressed by many cells and tissues and also by erythrocytes |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title