missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD173 Antibody (BRIC231), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-62120
This item is not returnable.
View return policy
Description
CD173 Monoclonal specifically detects CD173 in Human samples. It is validated for Flow Cytometry, Haemagglutination, Immunofluorescence.
Specifications
| CD173 | |
| Monoclonal | |
| 1 mg/mL | |
| Flow Cytometry 1:10-1:1000, Haemagglutination | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| NB100-62120 recognises the type 2 H blood group antigen, recently designated CD173. Active H substances in man, are expressed by many cells and tissues and also by erythrocytes | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Flow Cytometry, Hemagglutination Assay | |
| BRIC231 | |
| Unconjugated | |
| H2, Type 2 H blood group antigen | |
| HEL cell line | |
| 0.1 mg | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction