missing translation for 'onlineSavingsMsg'
Learn More

CD169 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen CD169
Concentration 0.49 mg/mL
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16876885
View Documents
Proteintech
55427-1-AP-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16866885
View Documents
Proteintech
55427-1-AP-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD169 are part of siglec family of membrane bound lectins (of the immunoglobulin superfamily) that bind sialic acid and mediate cell-cell interactions. Family members include sialoadhesin, CD22 and CD33. Sialoadhesin is a macrophage-specific cell adhesion molecule that is expressed on activated macrophages in chronic inflammation and in tumors.
Specifications

Specifications

CD169
Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Human
Q9BZZ2
6614
Peptide
Primary
-20°C
SIGLEC1
0.49 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
CD169, Sialoadhesin, SIGLEC 1, SIGLEC1, SN
SIGLEC1
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.