missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD169 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55427-1-AP-150UL
This item is not returnable.
View return policy
Description
CD169 are part of siglec family of membrane bound lectins (of the immunoglobulin superfamily) that bind sialic acid and mediate cell-cell interactions. Family members include sialoadhesin, CD22 and CD33. Sialoadhesin is a macrophage-specific cell adhesion molecule that is expressed on activated macrophages in chronic inflammation and in tumors.Specifications
| CD169 | |
| Polyclonal | |
| Unconjugated | |
| SIGLEC1 | |
| CD169, Sialoadhesin, SIGLEC 1, SIGLEC1, SN | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 6614 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.49 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9BZZ2 | |
| SIGLEC1 | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction