missing translation for 'onlineSavingsMsg'
Learn More

CD163 Antibody (CL10652), Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP3-07981-100ul

 View more versions of this product

Product Code. 18699757

  • £533.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

CD163 Monoclonal antibody specifically detects CD163 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD163
Monoclonal
Unconjugated
PBS, pH 7.2, containing 40% glycerol
Mouse
Protein A purified
RUO
Primary
Human, Rat
Purified
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
CL10652
Western Blot 1 μg/ml, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500
CD_antigen: CD163, CD163, CD163 molecule, Hemoglobin scavenger receptor, M130, macrophage-associated antigen, MM130, SCARI1, scavenger receptor cysteine-rich type 1 protein M130, sCD163, Soluble CD163
This antibody was developed against a recombinant protein corresponding to amino acids: ACKQLGCPTAVTAIGRVNASKGFGHIWLDSVSCQGHEPAVWQCKHHEWGKHYCNHNEDAGVTCSD
100 μg
Angiogenesis, Immune System Diseases, Immunology, Inflammation, Innate Immunity
9332
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.