missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD161 Antibody (14F1F11) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£155.00 - £467.00
Specifications
| Antigen | CD161 |
|---|---|
| Clone | 14F1F11 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1 - 5ug/ml, Immunohistochemistry 1:10 - 1:2000, Immunohistochemistry-Paraffin 5 ug/ml |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18111025
|
Novus Biologicals
NBP2-31359-0.025MG |
0.025 mg |
£155.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18108233
|
Novus Biologicals
NBP2-31359-0.1MG |
0.1 mg |
£467.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD161 Monoclonal specifically detects CD161 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| CD161 | |
| 1.0 mg/mL | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Signal Transduction | |
| PBS with No Preservative | |
| CD161, CD161 antigen, CLEC5BC-type lectin domain family 5 member B, HNKR-P1a, killer cell lectin-like receptor subfamily B member 1, killer cell lectin-like receptor subfamily B, member 1, Natural killer cell surface protein P1A, NKR-P1, NKR-P1AMGC138614, NKRP1ANKR | |
| KLRB1 | |
| IgG1 κ | |
| Protein G purified | |
| The antibody has not been tested in species samples other than human. |
| 14F1F11 | |
| Western Blot 1 - 5ug/ml, Immunohistochemistry 1:10 - 1:2000, Immunohistochemistry-Paraffin 5 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Q12918 | |
| 3820 | |
| Amino acids 68-225 of human CD161 were used as the immunogen for this antibody. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title