Learn More
CD161 Antibody (14F1F11) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-31359-0.1MG
 View more versions of this product
Product Code. 18108233
  • £467.00 / 0.10mg
Description
CD161 Monoclonal specifically detects CD161 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD161
Monoclonal
1.0 mg/mL
Western Blot 1 - 5ug/ml, Immunohistochemistry 1:10 - 1:2000, Immunohistochemistry-Paraffin 5 ug/ml
Q12918
KLRB1
Amino acids 68-225 of human CD161 were used as the immunogen for this antibody.
0.1 mg
Signal Transduction
3820
Human
Purified
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
14F1F11
Unconjugated
PBS with No Preservative
CD161, CD161 antigen, CLEC5BC-type lectin domain family 5 member B, HNKR-P1a, killer cell lectin-like receptor subfamily B member 1, killer cell lectin-like receptor subfamily B, member 1, Natural killer cell surface protein P1A, NKR-P1, NKR-P1AMGC138614, NKRP1ANKR
Mouse
Protein G purified
RUO
Primary
The antibody has not been tested in species samples other than human.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG1 κ