missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD160 Antibody (BY55), Novus Biologicals™
Mouse Monoclonal Antibody
£141.00 - £381.00
Specifications
| Antigen | CD160 |
|---|---|
| Clone | BY55 |
| Concentration | 0.1 mg/mL |
| Dilution | Flow Cytometry 1:10-1:50, Immunoprecipitation 1:10-1:500 |
| Applications | Flow Cytometry, Immunoprecipitation |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18282272
|
Novus Biologicals
NBP1-21435-0.025MG |
0.025 mg |
£141.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18286320
|
Novus Biologicals
NBP1-21435 |
0.1 mg |
£381.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD160 Monoclonal specifically detects CD160 in Human samples. It is validated for Flow Cytometry, Immunoprecipitation.Specifications
| CD160 | |
| 0.1 mg/mL | |
| Flow Cytometry, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Human | |
| BY55FLJ46513, CD160 antigenimmunoglobulin superfamily member, CD160 molecule, Natural killer cell receptor BY55 | |
| CD160 | |
| IgM | |
| IgM purified | |
| NBP1-21435 recognizes human CD160, a 27kD cell surface glycoprotein that was initially identified by clone BY55. The expression of CD160 is restricted to circulating NK cells, lymphocytes expressing T-cell receptor (TCR) gamma/delta and a small population of TCR alpha/beta positive T cells. In tissues, CD160 is expressed by intestinal intraepithelial lymphocytes. |
| BY55 | |
| Flow Cytometry 1:10-1:50, Immunoprecipitation 1:10-1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| O95971 | |
| 11126 | |
| Human NK cell line YT2C2 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title