missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD160 Antibody (BY55), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-21435-0.025MG
This item is not returnable.
View return policy
Description
CD160 Monoclonal specifically detects CD160 in Human samples. It is validated for Flow Cytometry, Immunoprecipitation.
Specifications
| CD160 | |
| Monoclonal | |
| 0.1 mg/mL | |
| Flow Cytometry 1:10-1:50, Immunoprecipitation 1:10-1:500 | |
| O95971 | |
| CD160 | |
| Human NK cell line YT2C2 | |
| 0.025 mg | |
| Innate Immunity, Signal Transduction | |
| 11126 | |
| Human | |
| Purified |
| Flow Cytometry, Immunoprecipitation | |
| BY55 | |
| Unconjugated | |
| PBS (pH 7.4) with 0.09% Sodium Azide | |
| BY55FLJ46513, CD160 antigenimmunoglobulin superfamily member, CD160 molecule, Natural killer cell receptor BY55 | |
| Mouse | |
| IgM purified | |
| RUO | |
| Primary | |
| NBP1-21435 recognizes human CD160, a 27kD cell surface glycoprotein that was initially identified by clone BY55. The expression of CD160 is restricted to circulating NK cells, lymphocytes expressing T-cell receptor (TCR) gamma/delta and a small population of TCR alpha/beta positive T cells. In tissues, CD160 is expressed by intestinal intraepithelial lymphocytes. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgM |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction