missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD16 Mouse anti-Baboon, Human, Rhesus, Brilliant Ultraviolet 496, Clone: 3G8, BD Horizon™
Mouse Monoclonal Antibody
£241.80 - £427.14
Specifications
| Antigen | CD16 |
|---|---|
| Clone | 3G8 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | Brilliant Ultraviolet 496 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17237994
|
BD Biosciences
612945 |
25 Tests |
£253.00 £241.80 / 25 tests Save £11.20 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17227994
|
BD Biosciences
612944 |
100 Tests |
£445.00 £427.14 / 100 tests Save £17.86 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD16 Monoclonal antibody specifically detects CD16 in Baboon, Cynomolgus, Human, Rhesus Ma samples. It is validated for Flow cytometry.Specifications
| CD16 | |
| Flow Cytometry | |
| Brilliant Ultraviolet 496 | |
| RUO | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Human polymorphonuclear leukocytes | |
| Affinity Purified |
| 3G8 | |
| Monoclonal | |
| Mouse | |
| Baboon, Human, Rhesus Monkey | |
| FcRIII; Fc-gamma RIII; FCG3; FCGR3; FCGRIII; FcγRIII; IGFR3 | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title