missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD16 Mouse anti-Baboon, Human, Rhesus, Brilliant Ultraviolet 496, Clone: 3G8, BD Horizon™
Mouse Monoclonal Antibody
Brand: BD Biosciences 612944
This item is not returnable.
View return policy
Description
CD16 Monoclonal antibody specifically detects CD16 in Baboon, Cynomolgus, Human, Rhesus Ma samples. It is validated for Flow cytometry.
Specifications
| CD16 | |
| Monoclonal | |
| Brilliant Ultraviolet 496 | |
| FcRIII; Fc-gamma RIII; FCG3; FCGR3; FCGRIII; FcγRIII; IGFR3 | |
| Human polymorphonuclear leukocytes | |
| 100 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| 3G8 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Baboon, Human, Rhesus Monkey |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction