Learn More
CD158b Mouse anti-Human, PE, Clone: CH-L, BD
Mouse Monoclonal Antibody
£125.74 - £220.33
Specifications
| Antigen | CD158b |
|---|---|
| Clone | CH-L |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | PE |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15881269
|
BD Biosciences
561723 |
25 Tests |
£131.00 £125.74 / Each Save £5.26 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15869778
|
BD Biosciences
559785 |
100 Tests |
£231.00 £220.33 / Each Save £10.67 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Reacts with a 50 - 58 kDa type I glycoprotein molecule of the killer cell inhibitory receptor (KIR) family of proteins. CD158b has also been known as GLI83, p50.2, or p58.2. CD158b is composed of two extracellular Ig-like domains. It is expressed on a subset of NK cells and it may also be present on rare T cells (either CD3/TCR αβ+, or CD3/TCR γδ+). CD158b regulates NK cell mediated cytolytic activity by interacting with HLA-C alleles (Cw 1, 3, 7 and 8).
Specifications
| CD158b | |
| Flow Cytometry | |
| PE | |
| RUO | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| CH-L | |
| Monoclonal | |
| Mouse | |
| Human | |
| IgG2b κ | |
| Affinity Purified |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.