missing translation for 'onlineSavingsMsg'
Learn More

CD158b Mouse anti-Human, PE, Clone: CH-L, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 559785

 View more versions of this product

Product Code. 15869778

  • ONLINE EXCLUSIVE PRICE
    £231.00
    £220.33 / Each
    Save £10.67
    4% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reacts with a 50 - 58 kDa type I glycoprotein molecule of the killer cell inhibitory receptor (KIR) family of proteins. CD158b has also been known as GLI83, p50.2, or p58.2. CD158b is composed of two extracellular Ig-like domains. It is expressed on a subset of NK cells and it may also be present on rare T cells (either CD3/TCR αβ+, or CD3/TCR γδ+). CD158b regulates NK cell mediated cytolytic activity by interacting with HLA-C alleles (Cw 1, 3, 7 and 8).

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD158b
Monoclonal
PE
Mouse
100 Tests
Immunology
Human
IgG2b κ
Flow Cytometry
CH-L
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Affinity Purified
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.