missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
CD155/PVR Rabbit anti-Mouse, Clone: 305, Novus Biologicals™
Rabbit Monoclonal Antibody
£309.00 - £392.00
Tekniske data
| Antigen | CD155/PVR |
|---|---|
| Clone | 305 |
| Dilution | Flow Cytometry 1:25-1:100, Immunocytochemistry/Immunofluorescence 1:50-1:1000 |
| Applications | Flow Cytometry, Immunofluorescence |
| Classification | Monoclonal |
| Produktkode | Brand | Quantity | Pris | Antal & tilgængelighed | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkode | Brand | Quantity | Pris | Antal & tilgængelighed | |||||
|
18224706
|
Novus Biologicals
NBP2-90501-50UL |
50 μg |
£309.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18297427
|
Novus Biologicals
NBP2-90501-100UL |
100 μg |
£392.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beskrivelse
CD155/PVR Monoclonal specifically detects CD155/PVR in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.Tekniske data
| CD155/PVR | |
| Flow Cytometry 1:25-1:100, Immunocytochemistry/Immunofluorescence 1:50-1:1000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| CD155, CD155 antigen, HVED, NECL5, NECL-5, nectin-like 5, Nectin-like protein 5, poliovirus receptor, PVSFLJ25946, Tage4 | |
| Recombinant Mouse CD155/PVR protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 305 | |
| Flow Cytometry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Virology Bacteria and Parasites | |
| 0.2 um filtered solution in PBS | |
| 5817 | |
| IgG | |
| Protein A purified |
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel